Tri(ethylene glycol) di-p-toluenesulfonate, 98%
Tri(ethylene glycol) di-p-toluenesulfonate, 98%
Tri(ethylene glycol) di-p-toluenesulfonate, 98%
Tri(ethylene glycol) di-p-toluenesulfonate, 98%
Thermo Scientific Chemicals

Tri(ethylene glycol) di-p-toluenesulfonate, 98%

CAS: 19249-03-7 | C20H26O8S2 | 458.54 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) B23279.09
または、製品番号B23279-09
価格(JPY)
-
見積もりを依頼する
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19249-03-7
IUPAC Name2-(2-{2-[(4-methylbenzenesulfonyl)oxy]ethoxy}ethoxy)ethyl 4-methylbenzene-1-sulfonate
Molecular FormulaC20H26O8S2
InChI KeyKCONMNWPRXAWKK-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOS(=O)(=O)C1=CC=C(C)C=C1
さらに表示
Appearance (Color)White
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)76.0-82.0?C
Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.

Solubility
Soluble in ethanol.

Notes
Incompatible with strong bases.
RUO – Research Use Only

General References:

  1. Tripathi, G.; Ramanathan, G. Structures and conformation of a benzo-12-crown-4 containing dipeptide. Pept. Sci. 2015, 104 (3), 148-155.
  2. Kazaylek, M.; Kocakaya, Ş. O.; Oral, E. V.; Çolak, M.; Karakaplan, M. Complexation and transport of amino acid esters and their salts with synthesised chiral novel aza crown ether derivatives. Supramol. Chem. 2014, 26 (5-6), 363-372.