2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
2,4-Dibromophenol, 98%
Thermo Scientific Chemicals

2,4-Dibromophenol, 98%

CAS: 615-58-7 | C6H4Br2O | 251.905 g/mol
製品番号(カタログ番号) B23394.22
または、製品番号B23394-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS615-58-7
IUPAC Name2,4-dibromophenol
Molecular FormulaC6H4Br2O
InChI KeyFAXWFCTVSHEODL-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1Br
さらに表示
FormCrystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only