Thermo Scientific Chemicals

1-Chlorohexadecane, 97%, Thermo Scientific Chemicals

Catalog number: B23406.AE
100 mL, Each
Thermo Scientific Chemicals

1-Chlorohexadecane, 97%, Thermo Scientific Chemicals

Catalog number: B23406.AE
100 mL, Each
Quantity
Catalog number: B23406.AE
also known as B23406-AE
Price (USD)
Price: 37.50
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4860-03-1
IUPAC Name
1-chlorohexadecane
Molecular Formula
C16H33Cl
InChI Key
CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCl
Identification (FTIR)
Conforms
Refractive Index
1.4475-1.4525 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%

Description

1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

Solubility
Soluble in ether. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text