2,2'-Dimethoxybiphenyl, 97%
2,2'-Dimethoxybiphenyl, 97%
2,2'-Dimethoxybiphenyl, 97%
2,2'-Dimethoxybiphenyl, 97%
Thermo Scientific Chemicals

2,2'-Dimethoxybiphenyl, 97%

CAS: 4877-93-4 | C14H14O2 | 214.26 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B23411.06
または、製品番号B23411-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4877-93-4
IUPAC Name2,2'-dimethoxy-1,1'-biphenyl
Molecular FormulaC14H14O2
InChI KeyVGMKUVCDINAAFC-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C1=CC=CC=C1OC
さらに表示
FormCrystals or powder or crystalline powder
Melting Point (clear melt)151.0-160.0?C
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Noriyuki Yonezawa; Shiro Miyata; Toshiaki Nakamura; Shinichiro Mori; Yuriko Ueha; Ryoichi Katakai. Synthesis of wholly aromatic polyketones using 2,2'-dimethoxybiphenyl as the acyl-acceptant monomer. Macromolecules. 1993, 26 (19), 5262-5263.
  2. N. P. Buu-Hoë; L. Petit; D. C. Thang. Orientation in Some Friedel—Crafts Acylations of 2,2'-Dimethoxybiphenyl, and the Cyclization of the Reaction Products. J. Org. Chem. 1964, 29 (3), 762-763.