1,8-Dichloroanthraquinone, 96%
1,8-Dichloroanthraquinone, 96%
1,8-Dichloroanthraquinone, 96%
1,8-Dichloroanthraquinone, 96%
Thermo Scientific Chemicals

1,8-Dichloroanthraquinone, 96%

CAS: 82-43-9 | C14H6Cl2O2 | 277.1 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) B23421.36
または、製品番号B23421-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS82-43-9
IUPAC Name1,8-dichloro-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H6Cl2O2
InChI KeyVBQNYYXVDQUKIU-UHFFFAOYSA-N
SMILESClC1=CC=CC2=C1C(=O)C1=C(Cl)C=CC=C1C2=O
さらに表示
Appearance (Color)Pale yellow to yellow
Assay (GC)≥95.0%
FormPowder
Melting Point (clear melt)199.0-208.0?C
1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. Kumao Hamanoue; Masaki Kimoto; Yōichi Kajiwara; Toshihiro Nakayama; Hiroshi Teranishi. Primary photoreduction of 1,8-dichloroanthraquinone by triethylamine studied by laser spectroscopy. Journal of Photochemistry. 1985, 31 (1), 143-147.
  2. Kumao Hamanoue; Kazuo Nakajima; Yōichi Kajiwara; Toshihiro Nakayama; Hiroshi Teranishi. Keller and Stephen G. Pyne. Observation of a new absorption due to a precursor of the lowest triplet state of 1,8-dichloroanthraquinone. Chemical Physics Letters. 1984, 110 (2), 178-182.