2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
Thermo Scientific Chemicals

2-Ethylhexanal, 96%

CAS: 123-05-7 | C8H16O | 128.215 g/mol
製品番号(カタログ番号) B23471.18
または、製品番号B23471-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123-05-7
IUPAC Name2-ethylhexanal
Molecular FormulaC8H16O
InChI KeyLGYNIFWIKSEESD-UHFFFAOYNA-N
SMILESCCCCC(CC)C=O
さらに表示
Free acid (titration)≤2.5% (non-U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4125-1.4185 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless
FormLiquid
さらに表示
2-Ethylhexanal is used as an intermediate for chemical synthesis and in the production of pharmaceuticals and odorous substances.  

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanal is used as an intermediate for chemical synthesis & in the production of disinfectants, pharmaceuticals and odorous substances.  

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Christel Lehtinen and Gösta Brunow. Epoxidation of Terminal Alkenes with Oxygen and 2-Ethyl Hexanal, without Added Catalyst or Solvent. Org. Proc. Res. Dev. 1999, 3, (2), 101-103.
  2. Christel Lehtinen and Gösta Brunow. Factors Affecting the Selectivity of Air Oxidation Of 2-Ethyhexanal, an α-Branched Aliphatic Aldehyde. Org. Proc. Res. Dev. 2000, 4 (6), 544-549.