4-Chlorobenzhydrol, 98%
4-Chlorobenzhydrol, 98%
4-Chlorobenzhydrol, 98%
Thermo Scientific Chemicals

4-Chlorobenzhydrol, 98%

CAS: 119-56-2 | C13H11ClO | 218.68 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B23691.14
または、製品番号B23691-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS119-56-2
IUPAC Name(4-chlorophenyl)(phenyl)methanol
Molecular FormulaC13H11ClO
InChI KeyAJYOOHCNOXWTKJ-UHFFFAOYNA-N
SMILESOC(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
さらに表示
Melting Point (clear melt)58.0-64.0?C
Assay (GC)≥97.5%
FormPowder or lumps
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only