3-Ethoxypropylamine, 99%
3-Ethoxypropylamine, 99%
3-Ethoxypropylamine, 99%
Thermo Scientific Chemicals

3-Ethoxypropylamine, 99%

CAS: 6291-85-6 | C5H13NO | 103.165 g/mol
製品番号(カタログ番号)数量
B23694.22
または、製品番号B23694-22
100 g
製品番号(カタログ番号) B23694.22
または、製品番号B23694-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6291-85-6
IUPAC Name3-ethoxypropan-1-amine
Molecular FormulaC5H13NO
InChI KeySOYBEXQHNURCGE-UHFFFAOYSA-N
SMILESCCOCCCN
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4180-1.4210 @ 20°C
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

Solubility
Miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
RUO – Research Use Only

General References:

  1. V. Baudel, F. Cazier, et al. Synthesis and modification of oxetane based oligomers with 3-ethoxypropylamine by focused microwave irradiation.Eur. Polym. J.,2002,38(3), 615-618.
  2. E. Uğuzdoğan, et al. A new temperature-sensitive polymer: Poly(ethoxypropylacrylamide).Eur. Polym. J.,2005,41(9), 2142-2149.