1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper
1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper
1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper
Thermo Scientific Chemicals

1,1,1-Trifluoro-4-iodobutane, 98+%, stab. with copper

CAS: 461-17-6 | C4H6F3I | 237.992 g/mol
製品番号(カタログ番号) B23811.03
または、製品番号B23811-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS461-17-6
IUPAC Name1,1,1-trifluoro-4-iodobutane
Molecular FormulaC4H6F3I
InChI KeyLNDGACQEAYKNOI-UHFFFAOYSA-N
SMILESFC(F)(F)CCCI
さらに表示
Assay (GC)> 98.0%
Formliquid
Appearance (Color)Clear, colourless to pale yellow or pale pink
Refractive Index1.4315 - 1.4355 @20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only