4-(1-Pyrrolidinyl)pyridine, 98%
4-(1-Pyrrolidinyl)pyridine, 98%
4-(1-Pyrrolidinyl)pyridine, 98%
Thermo Scientific Chemicals

4-(1-Pyrrolidinyl)pyridine, 98%

CAS: 2456-81-7 | C9H12N2 | 148.209 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B23989.14
または、製品番号B23989-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2456-81-7
IUPAC Name4-(pyrrolidin-1-yl)pyridin-1-ium
Molecular FormulaC9H13N2
InChI KeyRGUKYNXWOWSRET-UHFFFAOYSA-O
SMILESC1CCN(C1)C1=CC=[NH+]C=C1
さらに表示
Appearance (Color)White to pale cream to pale yellow
FormCrystals or powder or crystalline powder or lumps or fused solid and/or chunks
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only