1,3-Benzenedisulfonyl chloride, 97%
1,3-Benzenedisulfonyl chloride, 97%
1,3-Benzenedisulfonyl chloride, 97%
Thermo Scientific Chemicals

1,3-Benzenedisulfonyl chloride, 97%

CAS: 585-47-7 | C6H4Cl2O4S2 | 275.11 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B24044.22
または、製品番号B24044-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS585-47-7
IUPAC Namebenzene-1,3-disulfonyl dichloride
Molecular FormulaC6H4Cl2O4S2
InChI KeyALIQZUMMPOYCIS-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O
さらに表示
Appearance (Color)White to cream to pale brown
Assay (GC)≥96.0%
FormCrystals or crystalline powder
Melting Point (clear melt)56-62?C
Assay (HPLC)≥96.0%
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

Solubility
Soluble in DMSO and methanol. Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
RUO – Research Use Only

General References:

  1. Jens Kjær Jørgensen; Espen Ommundsen; Aage Stori; Keith Redford. Synthesis, characterisation and decomposition of 1,3-benzene disulfonyl azide; a cross-linking agent for polyolefins. Polymer. 2005, 46 (26), 12073-12080.
  2. Joerg C. Tiller; Raino Rieseler; Peter Berlin and Dieter Klemm. Stabilization of Activity of Oxidoreductases by Their Immobilization onto Special Functionalized Glass and Novel Aminocellulose Film Using Different Coupling Reagents. Biomacromolecules. 2002, 3 (5), 1021-1029.