2,3-Dimethoxybenzeneboronic acid, 97+%
2,3-Dimethoxybenzeneboronic acid, 97+%
2,3-Dimethoxybenzeneboronic acid, 97+%
Thermo Scientific Chemicals

2,3-Dimethoxybenzeneboronic acid, 97+%

CAS: 40972-86-9 | C8H11BO4 | 181.98 g/mol
製品番号(カタログ番号) B24125.06
または、製品番号B24125-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS40972-86-9
IUPAC Name(2,3-dimethoxyphenyl)boronic acid
Molecular FormulaC8H11BO4
InChI KeyVREWSCMOGIXMDQ-UHFFFAOYSA-N
SMILESCOC1=CC=CC(B(O)O)=C1OC
さらに表示
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.0%
Proton NMRConforms to structure
Assay (Aqueous acid-base Titration)≥97.0%
Appearance (Color)White to pale cream
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only