Thermo Scientific Chemicals

N,N-Dimethyldodecylamine, 90+%, Thermo Scientific Chemicals

Catalog number: B24145.18
50 g, Each
Thermo Scientific Chemicals

N,N-Dimethyldodecylamine, 90+%, Thermo Scientific Chemicals

Catalog number: B24145.18
50 g, Each
Quantity
Catalog number: B24145.18
also known as B24145-18
Price (USD)
Price: 29.50
Online price: 24.65
Your price:
Quantity
-

Chemical Identifiers

CAS
112-18-5
IUPAC Name
dodecyldimethylamine
Molecular Formula
C14H31N
InChI Key
YWFWDNVOPHGWMX-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCN(C)C
Comment
May contain trace amount of crystalline material believed to be N-oxide
Appearance (Color)
Clear to slightly hazy colorless
Assay (GC)
≥88.0%
Refractive Index
1.4335-1.4415 @ 20?C
Form
Liquid

Description

N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.

Solubility
Slightly soluble in water.

Notes
Air & moisture sensitive. Materials to avoid is copper, oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text