Thermo Scientific Chemicals

Maltitol, 97%, Thermo Scientific Chemicals

Catalog number: B24158.18
50 g, Each
Thermo Scientific Chemicals

Maltitol, 97%, Thermo Scientific Chemicals

Catalog number: B24158.18
50 g, Each
Quantity
Catalog number: B24158.18
also known as B24158-18
Price (USD)
Price: 49.00
Online price: 41.65
Your price:
Quantity
-

Chemical Identifiers

CAS
585-88-6
IUPAC Name
4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol
Molecular Formula
C12H24O11
InChI Key
VQHSOMBJVWLPSR-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO
Form
crystalline powder
Appearance (Color)
White
Optical Rotation
+107? ? 3? (c=10 in water)
Assay (unspecified)
>96.0%

Description

Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text