Methyl dichloroacetate, 99%
Methyl dichloroacetate, 99%
Methyl dichloroacetate, 99%
Thermo Scientific Chemicals

Methyl dichloroacetate, 99%

CAS: 116-54-1 | C3H4Cl2O2 | 142.963 g/mol
製品番号(カタログ番号) B24399.36
または、製品番号B24399-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS116-54-1
IUPAC Namemethyl 2,2-dichloroacetate
Molecular FormulaC3H4Cl2O2
InChI KeyHKMLRUAPIDAGIE-UHFFFAOYSA-N
SMILESCOC(=O)C(Cl)Cl
さらに表示
Water Content (Karl Fischer Titration)≤0.5%
Assay (GC)≥98.5%
Refractive Index1.4415-1.4445 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Methyl dichloroacetate may be used in the diastereoselctive synthesis of bicyclic chlorocyclopropane, as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate, as reagent during CrCl2-induced olefination of aldehydes. It is a reactant involved in nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes, azide-alkyl halide cycloaddition and atom transfer radical addition, Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines, three component reaction of arynes with cyclic ethers and active methines, catalytic radical haloalkylation for synthesis of neodysidenin and addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dichloroacetate may be used in the diastereoselctive synthesis of bicyclic chlorocyclopropane, as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate, as reagent during CrCl2-induced olefination of aldehydes. It is a reactant involved in nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes, azide-alkyl halide cycloaddition and atom transfer radical addition, Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines, three component reaction of arynes with cyclic ethers and active methines, catalytic radical haloalkylation for synthesis of neodysidenin and addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides.

Solubility
Soluble in water. (4 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure proper ventilation.
RUO – Research Use Only

General References:

  1. Ana Escribano.; Concepción Pedregal.; Rosario González.; Alejandro Fernández.; Keith Burton.; Gregory A Stephenson. Diastereoselective cyclopropanation of cyclic enones with methyl dichloroacetate anion. Tetrahedron. 2001, 57 (46),9423-9427 .
  2. Yoshiyuki Mido.; Masao Hashimoto. Conformational isomerism of methyl dichloroacetate. An infrared, Raman and 35Cl NQR study. J. Mol. Struct. 1985, 131 (1-2), 71-81 .