Thermo Scientific Chemicals

Tetrabromophthalic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: B24550.36
500 g, Each
Thermo Scientific Chemicals

Tetrabromophthalic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: B24550.36
500 g, Each
Quantity
Catalog number: B24550.36
also known as B24550-36
Price (USD)
Price: 176.00
Online price: 153.65
Your price:
Quantity
-

Chemical Identifiers

CAS
632-79-1
IUPAC Name
tetrabromo-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C8Br4O3
InChI Key
QHWKHLYUUZGSCW-UHFFFAOYSA-N
SMILES
BrC1=C2C(=O)OC(=O)C2=C(Br)C(Br)=C1Br
Elemental Analysis
Carbon C: 20.20-21.24% (Theoretical 20.72%)
Form
Powder
Melting Point (clear melt)
269-277?C
Appearance (Color)
White
Elemental Analysis
Bromine Br: 67.20-70.65% (Theoretical 68.93%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text