Thermo Scientific Chemicals

Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals

Catalog number: B24696.36
500 g, Each
Thermo Scientific Chemicals

Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals

Catalog number: B24696.36
500 g, Each
Quantity
Catalog number: B24696.36
also known as B24696-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
2582-30-1
IUPAC Name
N''-aminoguanidine; carbonic acid
Molecular Formula
C2H8N4O3
InChI Key
OTXHZHQQWQTQMW-UHFFFAOYSA-N
SMILES
OC(O)=O.NN=C(N)N
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms

Description

Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text