Thermo Scientific Chemicals

Ethyl cyclopropanecarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: B24870.22
100 g, Each
Thermo Scientific Chemicals

Ethyl cyclopropanecarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: B24870.22
100 g, Each
Quantity
Catalog number: B24870.22
also known as B24870-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
4606-07-9
IUPAC Name
ethyl cyclopropanecarboxylate
Molecular Formula
C6H10O2
InChI Key
LDDOSDVZPSGLFZ-UHFFFAOYSA-N
SMILES
CCOC(=O)C1CC1
Appearance (Color)
Clear colorless to pale blue
Form
Liquid
Refractive Index
1.4180-1.4220 @ 20°C
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclopropanecarboxylate is a reagent used in the addition of cyclopropane. It acts as annulation reagent for the conversion of aromatics to 2-methylindanones under Friedel-Crafts conditions.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text