5-Bromo-1,2,3-trimethoxybenzene, 97%
5-Bromo-1,2,3-trimethoxybenzene, 97%
5-Bromo-1,2,3-trimethoxybenzene, 97%
Thermo Scientific Chemicals

5-Bromo-1,2,3-trimethoxybenzene, 97%

CAS: 2675-79-8 | C9H11BrO3 | 247.09 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B24872.14
または、製品番号B24872-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2675-79-8
Appearance (Color)White to cream to yellow to orange to brown
FormPowder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)75.5-84.5°C
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only