2,6-Bis(p-tolyl)pyridine, 97%
2,6-Bis(p-tolyl)pyridine, 97%
2,6-Bis(p-tolyl)pyridine, 97%
Thermo Scientific Chemicals

2,6-Bis(p-tolyl)pyridine, 97%

CAS: 14435-88-2 | C19H17N | 259.352 g/mol
製品番号(カタログ番号) B24894.03
または、製品番号B24894-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS14435-88-2
化学物質名または材質2,6-Bis(p-tolyl)pyridine
EINECS番号238-409-3
式量259.35
健康被害1H315-H319-H335
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only