Cyclopentanone oxime, 97%
Cyclopentanone oxime, 97%
Cyclopentanone oxime, 97%
Thermo Scientific Chemicals

Cyclopentanone oxime, 97%

CAS: 1192-28-5 | C5H9NO | 99.13 g/mol
製品番号(カタログ番号) B24961.14
または、製品番号B24961-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1192-28-5
IUPAC NameN-cyclopentylidenehydroxylamine
Molecular FormulaC5H9NO
InChI KeyYGNXYFLJZILPEK-UHFFFAOYSA-N
SMILESON=C1CCCC1
さらに表示
FormCrystals or powder or crystalline powder
Melting Point (clear melt)52.0-61.0?C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only