5-Amino-4,6-dichloropyrimidine, 98+%
5-Amino-4,6-dichloropyrimidine, 98+%
5-Amino-4,6-dichloropyrimidine, 98+%
5-Amino-4,6-dichloropyrimidine, 98+%
Thermo Scientific Chemicals

5-Amino-4,6-dichloropyrimidine, 98+%

CAS: 5413-85-4 | C4H3Cl2N3 | 163.99 g/mol
製品番号(カタログ番号) B24985.06
または、製品番号B24985-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5413-85-4
IUPAC Name4,6-dichloropyrimidin-5-amine
Molecular FormulaC4H3Cl2N3
InChI KeyNIGDWBHWHVHOAD-UHFFFAOYSA-N
SMILESNC1=C(Cl)N=CN=C1Cl
さらに表示
Appearance (Color)White to cream
Melting Point (clear melt)141.0-148.0?C
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only