Thermo Scientific Chemicals

DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B25190.06
5 g, Each
Thermo Scientific Chemicals

DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: B25190.06
5 g, Each
Quantity
Catalog number: B25190.06
also known as B25190-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
132289-66-8
IUPAC Name
methyl 2-amino-2-(thiophen-2-yl)acetate hydrochloride
Molecular Formula
C7H10ClNO2S
InChI Key
NZEQIXANVTXFEP-UHFFFAOYNA-N
SMILES
Cl.COC(=O)C(N)C1=CC=CS1
Appearance (Color)
White to cream
Form
Powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text