Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (unspecified)≥95.0 to ≤105.0% (Calculated, dry wt. basis)
Assay (unspecified)Calcium ion: ≥15.0% (vendors CofA)
Assay (unspecified)Pyruvate anion: ≥60.0% (vendors CofA)
さらに表示