Thermo Scientific Chemicals

Coumalic acid, 97%, Thermo Scientific Chemicals

Catalog number: B25521.14
25 g, Each
Thermo Scientific Chemicals

Coumalic acid, 97%, Thermo Scientific Chemicals

Catalog number: B25521.14
25 g, Each
Quantity
Catalog number: B25521.14
also known as B25521-14
Price (USD)
Price: 121.00
Online price: 104.65
Your price:
Quantity
-

Chemical Identifiers

CAS
500-05-0
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point
ca. 200-215?C dec.
Form
Powder
Appearance (Color)
Pale yellow to cream or pale brown

Description

Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text