Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%
Thermo Scientific Chemicals

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%

CAS: 17159-80-7 | C9H16O3 | 172.22 g/mol
製品番号(カタログ番号) B25572.14
または、製品番号B25572-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS17159-80-7
Appearance (Color)Clear colorless to pale yellow to black
Assay (GC)≥96.0%
Refractive Index1.4635-1.4685 @ 20?C
FormLiquid
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Lixin Lang; Elaine Jagoda; Bernard Schmall; Bik-Kee Vuong; H. Richard Adams; David L. Nelson; Richard E. Carson and William C. Eckelman. Development of Fluorine-18-Labeled 5-HT1A Antagonists. J. Med. Chem. 1999, 42 (9), 1576-1586.
  2. Sebastian Imm; Sebastian Bähn; Prof. Dr. Min Zhang; Lorenz Neubert; Dr. Helfried Neumann; Dr. Florian Klasovsky; Dr. Jan Pfeffer; Dr. Thomas Haas; Prof. Dr. Matthias Beller. Improved Ruthenium-Catalyzed Amination of Alcohols with Ammonia: Synthesis of Diamines and Amino Esters. J. Med. Chem. 2011, 50 (33), 7599-7603.