Screw Vial Convenience Kit
Screw Vial Convenience Kit
Actual product may vary
Thermo Scientific™
Screw Vial Convenience Kit
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure Type
B7800-1-9A2 mLAmberSolid Top, Not Magnetic
B7990-24 mLClearScrew Cap, Open Top
B7990-38 mLClearScrew Cap, Open Top
B7800-1-92 mLClearSolid Top, Not Magnetic
B7990-1212 mLClearScrew Cap, Open Top
B7990-12A12 mLAmberScrew Cap, Open Top
B7990-416 mLClearScrew Cap, Open Top
Catalog number B7800-1-9A
Price (HKD)
-
Request A Quote
Volume (Metric):
2 mL
Color:
Amber
Closure Type:
Solid Top, Not Magnetic
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications
Certifications/ComplianceSpecification Certified
Closure ColorBlack
Closure MaterialPolypropylene
Closure Size8-425
Description8 mm Screw Amber Glass Vial Convenience Kit, Vial & Closure B7999-1A, B7815-9
Height (Metric)32 mm
Includes2mL amber glass vial, solid top PP screw cap
Product TypeAssembled, Unassembled Screw Vial Kit
Septa Size (Metric)8-425 mm
SeptumPTFE/Silicone liner
Closure TypeSolid Top, Not Magnetic
ColorAmber
Diameter (Metric) Outer12 mm
MaterialGlass
Pre-slitNo
QuantityPack of 100
Volume (Metric)2 mL
Unit SizePack of 100