Digitonin (5%) - FAQs
Digitonin (5%) - FAQs
View additional product information for Digitonin (5%) - FAQs (BN2006)
2 product FAQs found
I purchased Digitonin (5%) (Cat. No. BN2006) and the solution is whitish in color and seems to have precipitated. Is it okay to use?
If a precipitate forms in the 5% Digitonin solution, we recommend heating the solution at 95 degrees C for 5 mins and vortexing slowly to dissolve the precipitate. Cool to room temperature prior to use. The 5% Digitonin solution stays in solution at room temperature for up to 1 week.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.