MacConkey Broth Purple with Durham Tube
Thermo Scientific™

MacConkey Broth Purple with Durham Tube

Selectively detect and enumerate coliform bacteria in food, milk or water samples by observing lactose fermentation (colour change) and gas production, using the durham tube included with this broth.
製品番号(カタログ番号)充填重量
BO0347E10.0 to 10.3 g
製品番号(カタログ番号) BO0347E
価格(JPY)
-
充填重量:
10.0 to 10.3 g
Selectively detect and enumerate coliform bacteria in food, milk or water samples by observing lactose fermentation (colour change) and gas production, using MacConkey Broth Purple with Durham Tube
仕様
色Purple
充填重量10.0 to 10.3 g
フォーマットUniversal bottle
パッケージングタイプ24 Universal Bottles with screw cap closures in a box
数量24/Pk.
品質保持期間52 Weeks
製造元United Kingdom
製品タイプMicrobiology Media
Unit SizeEach
組成および保存条件
2°C to 25°C