Tryptone Soya Broth(ACC-EP/USP)1% Tween
Thermo Scientific™

Tryptone Soya Broth(ACC-EP/USP)1% Tween

Recover and enumerate microorganisms from pharmaceutical products or surfaces, neutralizing residual disinfectants for accurate sterility testing.
製品番号(カタログ番号)充填重量
BO1011N160 to 164 g
製品番号(カタログ番号) BO1011N
価格(JPY)
-
充填重量:
160 to 164 g
Tryptone Soya Broth (ACC-EP/USP)1%Tween is for recovery and enumeration of microorganisms from pharmaceutical products or surfaces, neutralizing residual disinfectants for accurate sterility testing.
仕様
色Straw
充填重量160 to 164 g
フォーマット250 mL DIN Bottle
パッケージングタイプ10 DIN Bottles/Box
数量10/Pk.
品質保持期間52 Weeks
製造元United Kingdom
製品タイプMicrobiology Media
Unit SizeEach
組成および保存条件
2°C to 25°C