Choice™ Polypropylene (PP) Syringe Filters

Choice™ Polypropylene (PP) Syringe Filters

Use Choice Polypropylene Syringe Filters as an economic solution to filtration of biological samples, and for filtration of aggressive organic solutions.

製品番号(カタログ番号)孔径直径(メートル法)
CH4513-PP0.45 μm13 mm
CH2225-PP0.2 μm25 mm
CH4525-PP0.45 μm25 mm
CH2213-PP0.2 μm13 mm
製品番号(カタログ番号) CH4513-PP
価格(JPY)
-
孔径:
0.45 μm
直径(メートル法):
13 mm

Thermo Scientific™ Choice™ Polypropylene Syringe Filters are a good choice for filtration of biological samples, and for filtration of aggressive organic solutions. They provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size.

Adding a syringe filtration step prior to chromatographic injection not only helps to ensure more consistent and reliable results, it also helps protect instruments and prolong column life. Economic Choice Syringe Filters are available in a variety of sizes and membrane types to meet any application need.

Specifications
  • Membrane: Hydrophilic polypropylene
  • Maximum operating temperature: 110 °C
  • Housing: Medical grade, virgin polypropylene
Applications
  • Protein- or peptide-based assays
  • General HPLC analysis
Chemical Incompatibilities
  • Hexane, toluene, benzene, limited resistance to dichloromethane and chloroform
仕様
直径(メートル法)13 mm
使用対象(アプリケーション)HPLC, SPE
ハウジング材質Solvent Resistent, Low-Extractable Polypropylene
プレフィルターNo
圧力定格90 psi
製品ラインChoice
数量100/Pk.
タイプSyringe Filter
ディスポーザブルSingle-use
材料Polypropylene
材料(膜)Polypropylene (Hydrophilic)
孔径0.45 μm
無菌性Non-sterile
Unit SizePack of 100