Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Fisher Chemical™
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C
数量:
1 L
2.5 L
パッケージ:
Amber Glass Bottle
製品番号(カタログ番号) D-1852-15
または、製品番号D/1852/15
価格(JPY)
-
お問い合わせください ›
数量:
1 L
パッケージ:
Amber Glass Bottle
一括またはカスタム形式をリクエストする
仕様
沸点39°C
CAS75-09-2
ChEBICHEBI:15767
Colorless
式量84.93g/mol
さらに表示
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C