DHX™ Heat Exchanger BioProcess Container (BPC), ASI™ 26/77 film, ReadyMate connections, four 6-L BPCs
Thermo Scientific™

DHX™ Heat Exchanger BioProcess Container (BPC), ASI™ 26/77 film, ReadyMate connections, four 6-L BPCs

The Thermo Scientific™ DHX™ Heat Exchanger Single-Use BioProcess Container (BPC) is a 2D system consisting of four 6 L BPCs,詳細を見る
製品番号(カタログ番号)数量
DX00009I
または、製品番号DX00009-I
4 x 6 L
製品番号(カタログ番号) DX00009I
または、製品番号DX00009-I
価格(JPY)
-
数量:
4 x 6 L
The Thermo Scientific™ DHX™ Heat Exchanger Single-Use BioProcess Container (BPC) is a 2D system consisting of four 6 L BPCs, without drain. It is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, with interconnecting TPE tubing and ReadyMate DAC end connections. The DHX Heat Exchanger and DHX BPC each features a serpentine pattern, creating a sterile fluid path for the process fluid. The BPC fits tightly between the stainless steel DHX Heat Exchanger plate system, and after loading, no further operator interaction is required. The customer has the option of utilizing up to 4 BPCs in the DHX unit.

Features
• Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks
• Provides sterile processing environment without the cost and time of clean-in-place or steam-in-place sterilization (CIP/SIP)
• Minimizes bioburden by using the DHX Heat Exchanger unit and DHX BPCs to address weak points in a CIP system or sanitation process (typically required for stainless steel equipment)
• Reduces maintenance and life cycle cost due to its unique design
• Increases volume capacity via parallel connection between DHX units

Applications
• cGMP commercial and clinical biotherapeutics, vaccines, and other biologic processes
• Upstream applications, including media hold, harvest cooling, and harvest hold
• Downstream applications, including harvest hold, buffers, protein purification, and bulk drug substance

仕様
寸法Each BPC: 62.2 cm x 61.9 cm (24.5 in. x 24.4 in.)
フィルム詳細ASI™ 26/77
使用対象 (装置)DHX1001
材料Polyethylene
外部包装Two 4-mil. poly bags
ポート詳細ReadyMate DAC connectors
製品ラインDHX
保護用内部包装Protective material around rough areas
数量4 x 6 L
出荷条件Room Temperature
滅菌方法Gamma Irradiated
容量(メートル法)24 L
Tubing StyleTPE
Unit SizeEach
組成および保存条件
Store at room temperature.