Fluorescein Di-β-D-Glucuronide (FDGlcU) - FAQs
View additional product information for Fluorescein Di-β-D-Glucuronide (FDGlcU) - FAQs (F2915)
2 product FAQs found
Can I make a higher stock solution of Fluorescein Di-β-D-Glucuronide (FDGlcU) (Cat. No. F2915) e.g., 50 mM in PBS?
Unfortunately, we do not have data on the upper limit of solubility for this reagent in PBS. This is something you would need to test empirically.br/>
Find additional tips, troubleshooting help, and resources within our Flow Cytometry Support Center.
What is the molecular weight of Fluorescein Di-β-D-Glucuronide (FDGlcU) (Cat. No. F2915)?
The molecular weight of the above product is 684.56 g/mol.br/>
Find additional tips, troubleshooting help, and resources within our Flow Cytometry Support Center.