FluoSpheres™ Carboxylate-Modified Microspheres
Invitrogen™

FluoSpheres™ Carboxylate-Modified Microspheres

Achieve the brightest fluorescence with Carboxylate-Modified FluoSphere Microspheres, available in different colors and particle sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)ColorExcitation/EmissionQuantity
F88110.2 μmYellow-Green505/515 nm10 mL
F88030.1 μmYellow-Green505/515 nm10 mL
F88161.0 μmCrimson625/645 nm2 mL
F88231.0 μmYellow-Green505/515 nm10 mL
F88010.1 μmRed580/605 nm10 mL
F88070.2 μmDark Red660/680 nm2 mL
F107200.04 μmYellow-Green, Orange, Red, Dark Red505/515, 540/560, 580/605, 660/680 nm1 mL/each
F208810.2 μmOrange365/610 nm2 mL
F8783
also known as F-8783
0.02 μmDark Red660/680 nm2 mL
F87860.02 μmRed580/605 nm10 mL
F8795
also known as F-8795
0.04 μmYellow-Green505/515 nm1 mL
F88130.5 μmYellow-Green505/515 nm10 mL
F88201.0 μmOrange540/560 nm10 mL
F88252.0 μmNile Red535/575 nm2 mL
F88272.0 μmYellow-Green505/515 nm2 mL
F8781
also known as F-8781
0.02 μmBlue365/415 nm10 mL
F8782
also known as F-8782
0.02 μmCrimson625/645 nm2 mL
F87840.02 μmNile Red535/575 nm10 mL
F87870.02 μmYellow-Green505/515 nm10 mL
F8789
also known as F-8789
0.04 μmDark Red660/680 nm1 mL
F8792
also known as F-8792
0.04 μmOrange540/560 nm1 mL
F87930.04 μmRed580/605 nm1 mL
F87940.04 μmRed-Orange565/580 nm1 mL
F87970.1 μmBlue350/440 nm10 mL
F87990.1 μmInfrared715/755 nm1 mL
F8800
also known as F-8800
0.1 μmOrange540/560 nm10 mL
F88050.2 μmBlue365/415 nm10 mL
F88060.2 μmCrimson625/645 nm2 mL
F88090.2 μmOrange540/560 nm10 mL
F88100.2 μmRed580/605 nm10 mL
F88120.5 μmRed580/605 nm10 mL
F88141.0 μmBlue365/415 nm10 mL
F88151.0 μmBlue350/440 nm10 mL
F88191.0 μmNile Red535/575 nm10 mL
F88211.0 μmRed580/605 nm10 mL
F8824
also known as F-8824
2.0 μmBlue365/415 nm2 mL
F88262.0 μmRed580/605 nm2 mL
Catalog number F8811
Price (USD)
536.00
Each
Diameter (Metric):
0.2 μm
Color:
Yellow-Green
Excitation/Emission:
505/515 nm
Quantity:
10 mL
Request bulk or custom format
Price (USD)
536.00
Each

Easily perform flow cytometry, microscopy, HTS, HCS, immunoassay, and other laboratory applications using our extensive selection of FluoSpheres Carboxylate-Modified Microspheres. FluoSphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications. FluoSphere microspheres are loaded with proprietary fluorescent dyes, making them the brightest microspheres available.

Visualize the brightest fluorescence for laboratoy applications including fluorescence microscopy, flow cytometry, HTS, HCS, and cell tracing with our Carboxylate-Modified FluoSphere Microspheres, which are manufactured from polystyrene microspheres and loaded with different proprietary dyes. Using specialized staining methods enables all of the fluorescent dye molecules to be contained inside each polystyrene microsphere instead of on the bead's surface. This protective environment within the bead shields the dye from detrimental environmental effects, such as photobleaching. Our carboxylate-modified microspheres are coated with a hydrophilic polymer containing multiple carboxylic acids for covalent attachment of ligands. A range of particle sizes is available for different research uses and experiments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation/Emission505/515 nm
Product LineFLUOSPHERES
Quantity10 mL
Surface ModificationCarboxylate
ColorYellow-Green
Diameter (Metric)0.2 μm
For Use With (Application)Fluorescence Microscopy
MaterialPolystyrene
Product TypeCarboxylate-Modified Microsphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

I have some FluoSpheres polystyrene microspheres, with 20 nm diameter. They are aggregating a lot. What can I do about it?

The smaller the microspheres, the greater the propensity to aggregate. But the aggregation is not irreversible. Sonicate in a bath sonicator or vortex to disperse, just prior to use. You can also add a small concentration of Tween-20 or Triton X-100 (unless you are using them in a live-cell system).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I sonicated my 2.0 µm carboxylate-modified microspheres, as recommended, but saw foaming (bubbles) on top of the solution. Should I be concerned?

Use of a bath sonicator is recommended to help break up any aggregated microspheres. The foaming is from Tween-20, which is in the stock solution to help prevent aggregation. It is normal and expected to see bubbles from this. Do not use a probe sonicator, which would cause damage to the microspheres (as well as much more bubbling).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When does the warranty guarantee expire for FluoSpheres Carboxylated-Modified Microspheres?

FluoSpheres Carboxylate-Modified Microspheres (Cat. Nos. F8783, F8786, F8801, F8807, F8811, F8803, F8816, F8823, F20881, F10720) have a 1-year warranty guarantee, unless otherwise indicated on the Certificate of Analysis (COA).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (10)

Citations & References
Abstract
Thermal fluctuations of grafted microtubules provide evidence of a length-dependent persistence length.
Authors:Pampaloni F, Lattanzi G, Jonás A, Surrey T, Frey E, Florin EL
Journal:Proc Natl Acad Sci U S A
PubMed ID:16801537
'Microtubules are hollow cylindrical structures that constitute one of the three major classes of cytoskeletal filaments. On the mesoscopic length scale of a cell, their material properties are characterized by a single stiffness parameter, the persistence length l(p). Its value, in general, depends on the microscopic interactions between the constituent ... More
Gridlock signalling pathway fashions the first embryonic artery.
Authors:Zhong TP, Childs S, Leu JP, Fishman MC
Journal:Nature
PubMed ID:11700560
'Arteries and veins are morphologically, functionally and molecularly very different, but how this distinction is established during vasculogenesis is unknown. Here we show, by lineage tracking in zebrafish embryos, that angioblast precursors for the trunk artery and vein are spatially mixed in the lateral posterior mesoderm. Progeny of each angioblast, ... More
Why molecules move along a temperature gradient.
Authors:Duhr S, Braun D
Journal:Proc Natl Acad Sci U S A
PubMed ID:17164337
'Molecules drift along temperature gradients, an effect called thermophoresis, the Soret effect, or thermodiffusion. In liquids, its theoretical foundation is the subject of a long-standing debate. By using an all-optical microfluidic fluorescence method, we present experimental results for DNA and polystyrene beads over a large range of particle sizes, salt ... More
Rapid transport of large polymeric nanoparticles in fresh undiluted human mucus.
Authors:Lai SK, O'Hanlon DE, Harrold S, Man ST, Wang YY, Cone R, Hanes J
Journal:Proc Natl Acad Sci U S A
PubMed ID:17244708
'Nanoparticles larger than the reported mesh-pore size range (10-200 nm) in mucus have been thought to be much too large to undergo rapid diffusional transport through mucus barriers. However, large nanoparticles are preferred for higher drug encapsulation efficiency and the ability to provide sustained delivery of a wider array of ... More
Real-time multiple particle tracking of gene nanocarriers in complex biological environments.
Authors:Lai SK, Hanes J,
Journal:Methods Mol Biol
PubMed ID:18470640
'Complex biological fluids, such as the vast and molecularly crowded cell cytoplasm and the highly viscoelastic mucus that protects many entry ways to the body, pose significant barriers to efficient gene delivery. Understanding the dynamics of gene carriers in such environments allows insight that leads to rational improvements in gene ... More