(R)-(+)-1,1'-Bi(2-naphthylamine), 97%
(R)-(+)-1,1'-Bi(2-naphthylamine), 97%
(R)-(+)-1,1'-Bi(2-naphthylamine), 97%
Thermo Scientific Chemicals

(R)-(+)-1,1'-Bi(2-naphthylamine), 97%

CAS: 18741-85-0 | C20H16N2 | 284.36 g/mol
製品番号(カタログ番号) H25785.03
または、製品番号H25785-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18741-85-0
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Optical Rotation+152.5? ±3? (c=1, pyridine)
FormPowder
(R)-(+)-1,1'-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1,1′-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kurt Mislow; Paul Grasemann. Notes - Absolute Configuration of 1,1'-Bi-2-naphthylamine. J. Org. Chem. 1958, 23 (12), 2027-2028.
  2. Shane G. Telfer; Tomohiro Sato; Takunori Harada; Reiko Kuroda; Julie Lefebvre and Daniel B. Leznoff. Mono- and Dinuclear Complexes of Chiral Tri- and Tetradentate Schiff-Base Ligands Derived from 1,1‘-Binaphthyl-2,2‘-diamine. Inorg. Chem. 2004, 43 (20), 6168-6176.