4-Nitro-alpha-toluenesulfonyl chloride, 97%
4-Nitro-alpha-toluenesulfonyl chloride, 97%
4-Nitro-alpha-toluenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Nitro-alpha-toluenesulfonyl chloride, 97%

CAS: 4025-75-6 | C7H6ClNO4S | 235.638 g/mol
製品番号(カタログ番号) H25794.06
または、製品番号H25794-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4025-75-6
IUPAC Name(4-nitrophenyl)methanesulfonyl chloride
Molecular FormulaC7H6ClNO4S
InChI KeyCAXRPEAFHWDFTD-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CS(Cl)(=O)=O)C=C1
さらに表示
FormPowder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)88-94°C
It is used as an intermediate in organic syntheses, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic syntheses, pharmaceuticals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
RUO – Research Use Only
Liuqun Gu; Yongyong Wu; Yazhu Zhang; Gang Zhao. A new class of efficient poly(ethylene-glycol)-supported catalyst based on proline for the asymmetric Michael addition of ketones to nitrostyrenes. Journal of Molecular Catalysis A: Chemical. 2007, 263 (1-2), 186-194.