D-Phenylalanine methyl ester hydrochloride, 98%
D-Phenylalanine methyl ester hydrochloride, 98%
D-Phenylalanine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

D-Phenylalanine methyl ester hydrochloride, 98%

CAS: 13033-84-6 | C10H14ClNO2 | 215.68 g/mol
製品番号(カタログ番号) H25920.14
または、製品番号H25920-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13033-84-6
IUPAC Namemethyl (2R)-2-amino-3-phenylpropanoate hydrochloride
Molecular FormulaC10H14ClNO2
InChI KeySWVMLNPDTIFDDY-SBSPUUFOSA-N
SMILESCl.COC(=O)[C@H](N)CC1=CC=CC=C1
さらに表示
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormPowder
Assay (Freebase GC)≥97.5%
Optical Rotation-37.0 ± 1.0° (c=2 in ethanol)
Benzhydrylidene Aza-Glycinyl-D-Phenylalanine Methyl Ester was synthesized from D-phenylalanine methyl ester hydrochloride. Used in the conversion of amino acids into n-menthyloxycarbonyl alkyl ester derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydrylidene Aza-Glycinyl-D-Phenylalanine Methyl Ester was synthesized from D-phenylalanine methyl ester hydrochloride. Used in the conversion of amino acids into n-menthyloxycarbonyl alkyl ester derivatives.

Solubility
Soluble in Ethanol and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only

General References:

  1. N. Domergue,; M. Pugniere,; A. Previero. One-Step Conversion of Amino Acids into N-Menthyloxycarbonyl Alkyl Ester Derivatives for Chiral Gas Chromatography. Analytical Biochemistry 1993, 214 (2), 420-425.
  2. Sano Shigeki; Nakao Michiyasu; Takeyasu Masanori; Kitaike Syuji; Yoshioka Yasuko; Nagao Yoshimitsu. Use Of Diketopiperazines For Determining Absolute Configurations Of Α-Substituted Serines By 1h-Nmr Spectroscopy. Journal name. 2009, 79 (1), 781-789.