Thermo Scientific Chemicals

Albendazole, 98+%, Thermo Scientific Chemicals

Catalog number: H25925.09
10 g, Each
Thermo Scientific Chemicals

Albendazole, 98+%, Thermo Scientific Chemicals

Catalog number: H25925.09
10 g, Each
Quantity
Catalog number: H25925.09
also known as H25925-09
Price (USD)
Price: 76.90
Online price: 65.65
Your price:
Quantity
-

Chemical Identifiers

CAS
54965-21-8
IUPAC Name
methyl N-[6-(propylsulfanyl)-1H-1,3-benzodiazol-2-yl]carbamate
Molecular Formula
C12H15N3O2S
InChI Key
HXHWSAZORRCQMX-UHFFFAOYSA-N
SMILES
CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1
Form
Powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text