5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H26020.06
または、製品番号H26020-06
5 g
製品番号(カタログ番号) H26020.06
または、製品番号H26020-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS889939-48-4
IUPAC Name5-chloro-2-methoxy-4-methylbenzene-1-sulfonyl chloride
Molecular FormulaC8H8Cl2O3S
InChI KeyBJRJHJZVSWZXIP-UHFFFAOYSA-N
SMILESCOC1=C(C=C(Cl)C(C)=C1)S(Cl)(=O)=O
さらに表示
Melting Point (clear melt)123.0-133.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only