2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals
2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals
2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H26030.031 g
製品番号(カタログ番号) H26030.03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS373-96-6
IUPAC Name2,2-difluoropropanoic acid
Molecular FormulaC3H4F2O2
InChI KeyPMWGIVRHUIAIII-UHFFFAOYSA-N
SMILESCC(F)(F)C(O)=O
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)35-47°C
Appearance (Color)White to pale yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only