Thermo Scientific Chemicals

1-(Trifluoromethyl)cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26055.03
1 g, Each
Thermo Scientific Chemicals

1-(Trifluoromethyl)cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26055.03
1 g, Each
Quantity
Catalog number: H26055.03
also known as H26055-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
277756-44-2
IUPAC Name
1-(trifluoromethyl)cyclopentane-1-carboxylic acid
Molecular Formula
C7H9F3O2
InChI Key
DGQRYPPBAJNZFZ-UHFFFAOYSA-N
SMILES
OC(=O)C1(CCCC1)C(F)(F)F
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Melting Point (clear melt)
33.0-42.0?C

Description

1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text