5-Fluoro-2-nitrophenylacetonitrile, 99%, Thermo Scientific Chemicals
5-Fluoro-2-nitrophenylacetonitrile, 99%, Thermo Scientific Chemicals
5-Fluoro-2-nitrophenylacetonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-nitrophenylacetonitrile, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H26125.03
または、製品番号H26125-03
1 g
製品番号(カタログ番号) H26125.03
または、製品番号H26125-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3456-75-5
IUPAC Name2-(5-fluoro-2-nitrophenyl)acetonitrile
Molecular FormulaC8H5FN2O2
InChI KeyYETOJTGGLXHUCS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1CC#N
さらに表示
FormCrystals or powder or crystalline powder
Melting Point (clear melt)67.0-73.0?C
Assay (GC)≥98.5%
Appearance (Color)White to pale cream to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only