2-Chloro-5-methylaniline, 98%
2-Chloro-5-methylaniline, 98%
2-Chloro-5-methylaniline, 98%
Thermo Scientific Chemicals

2-Chloro-5-methylaniline, 98%

CAS: 95-81-8 | C7H8ClN | 141.60 g/mol
製品番号(カタログ番号) H26209.06
または、製品番号H26209-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-81-8
IUPAC Name2-chloro-5-methylaniline
Molecular FormulaC7H8ClN
InChI KeyHPSCXFOQUFPEPE-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C(N)=C1
さらに表示
Melting Point (clear melt)28.0-35.0?C
FormCrystals or powder or crystalline powder or fused solid or flakes
Appearance (Color)White to cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only