4-(Trimethylsilyl)diphenylacetylene, 99%
4-(Trimethylsilyl)diphenylacetylene, 99%
4-(Trimethylsilyl)diphenylacetylene, 99%
4-(Trimethylsilyl)diphenylacetylene, 99%
Thermo Scientific Chemicals

4-(Trimethylsilyl)diphenylacetylene, 99%

CAS: 136459-72-8 | C17H18Si | 250.416 g/mol
製品番号(カタログ番号) H26300.06
または、製品番号H26300-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS136459-72-8
IUPAC Nametrimethyl[4-(2-phenylethynyl)phenyl]silane
Molecular FormulaC17H18Si
InChI KeyUZRJWRFLYPENFH-UHFFFAOYSA-N
SMILESC[Si](C)(C)C1=CC=C(C=C1)C#CC1=CC=CC=C1
さらに表示
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)35.0-42.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only