4-(4-Morpholinylmethyl)benzoic acid, 97%, Thermo Scientific Chemicals
4-(4-Morpholinylmethyl)benzoic acid, 97%, Thermo Scientific Chemicals
4-(4-Morpholinylmethyl)benzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(4-Morpholinylmethyl)benzoic acid, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
H26364.03
または、製品番号H26364-03
1 g
製品番号(カタログ番号) H26364.03
または、製品番号H26364-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS62642-62-0
IUPAC Name4-[(4-carboxylatophenyl)methyl]morpholin-4-ium
Molecular FormulaC12H15NO3
InChI KeyQYBXZYYECZFQRX-UHFFFAOYSA-N
SMILES[O-]C(=O)C1=CC=C(C[NH+]2CCOCC2)C=C1
さらに表示
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)177.0-186.0?C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only