Thermo Scientific Chemicals

Hypericin, 98%, Thermo Scientific Chemicals

Catalog number: H26425.MC
100 mg, Each
Thermo Scientific Chemicals

Hypericin, 98%, Thermo Scientific Chemicals

Catalog number: H26425.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H26425.MC
also known as H26425-MC
Price (USD)
Price: 1,716.00
Online price: 1,458.65
Your price:
Quantity
-

Chemical Identifiers

CAS
548-04-9
IUPAC Name
7,11,13,16,18,22-hexahydroxy-5,24-dimethyloctacyclo[13.11.1.1²,¹⁰.0³,⁸.0⁴,²⁵.0¹⁹,²⁷.0²¹,²⁶.0¹⁴,²⁸]octacosa-1(27),2(28),3,5,7,10,12,14,16,18,21,23,25-tridecaene-9,20-dione
Molecular Formula
C30H16O8
InChI Key
BTXNYTINYBABQR-UHFFFAOYSA-N
SMILES
CC1=CC(O)=C2C(=O)C3=C(O)C=C(O)C4=C5C(O)=CC(O)=C6C(=O)C7=C(O)C=C(C)C8=C7C(=C56)C(C2=C18)=C34
Assay from Supplier's CofA
≥97.5%
Appearance (Color)
Dark brown to black
Form
Powder

Description

Inhibitor of protein kinase C; induces photosynthesized apoptosisHypercin acts as an antibiotic and antiviral. It is involved in the inhibition of enzyme dopamine beta-hydroxylase action. It also exhibits an anti-HIV and anticancer drug. Further, it is useful in photodynamic therapy. In addition to this, it serves as an inhibitor of protein kinase C which induces photosynthesized apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hypericin is an anthraquinone derivative found in the yellow flower of Hypericum perforatum (also known as St. John's wort)
  • This compound has the ability to inhibit protein kinase C, inducing photosynthesized apoptosis

Application

  • It shows antibiotic and antiviral activities
  • The antidepressant activity of this compound could be related to its ability to inhibit the neuronal uptake of serotonin, norepinephrine, dopamine, gamma-aminobutyric acid (GABA), and L-glutamate
  • Since this compound is able to inhibit the assembly and shedding of virus particles in infected cells, hypericin may also prevent the replication of encapsulated viruses
  • It has been described that this compound can also exhibit anti-HIV and anticancer activities
  • Hypericin also displays an effective phototoxic activity and is able to trigger apoptotic pathways, resulting in the formation of reactive oxygen species
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text