3-Chloro-2-nitrobenzoic acid, 97%
3-Chloro-2-nitrobenzoic acid, 97%
3-Chloro-2-nitrobenzoic acid, 97%
Thermo Scientific Chemicals

3-Chloro-2-nitrobenzoic acid, 97%

CAS: 4771-47-5 | C7H4ClNO4 | 201.562 g/mol
製品番号(カタログ番号) H26467.06
または、製品番号H26467-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4771-47-5
IUPAC Name3-chloro-2-nitrobenzoate
Molecular FormulaC7H3ClNO4
InChI KeyVCHSXYHBMFKRBK-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CC(Cl)=C1[N+]([O-])=O
さらに表示
Appearance (Color)White
Assay (Silylated GC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)233.0-242.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only