2,3-Difluoro-6-methoxybenzoic acid, 97%
2,3-Difluoro-6-methoxybenzoic acid, 97%
2,3-Difluoro-6-methoxybenzoic acid, 97%
Thermo Scientific Chemicals

2,3-Difluoro-6-methoxybenzoic acid, 97%

CAS: 773873-26-0 | C8H6F2O3 | 188.13 g/mol
製品番号(カタログ番号) H26830.09
または、製品番号H26830-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS773873-26-0
IUPAC Name2,3-difluoro-6-methoxybenzoic acid
Molecular FormulaC8H6F2O3
InChI KeyMMTKYWQMCSZCGW-UHFFFAOYSA-N
SMILESCOC1=CC=C(F)C(F)=C1C(O)=O
さらに表示
Assay (Silylated GC)≥96.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)123.5-129.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only