1-Ethyl-3-methylimidazolium dicyanamide, 98%
1-Ethyl-3-methylimidazolium dicyanamide, 98%
1-Ethyl-3-methylimidazolium dicyanamide, 98%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium dicyanamide, 98%

CAS: 370865-89-7 | C8H11N5 | 177.211 g/mol
製品番号(カタログ番号) H26901.18
または、製品番号H26901-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS370865-89-7
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium; [(cyanoimino)methylidene]azanide
Molecular FormulaC8H11N5
InChI KeyMKHFCTXNDRMIDR-UHFFFAOYSA-N
SMILES[N-]=C=NC#N.CCN1C=C[N+](C)=C1
さらに表示
Appearance (Color)Clear colorless to yellow to orange-brown
Assay from Suppliers CofA≥97.5%
Refractive Index1.5130-1.5170 @ 20°C
FormLiquid
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Soluble in water. Soluble in solvents like methanol, acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Joan Fuller,; Richard T. Carlin,; Hugh C. De Long,; Dustin Haworth. Structure of 1-ethyl-3-methylimidazolium hexafluorophosphate: model for room temperature molten salts. J. Chem. Soc., Chem. Commun.1994 , (3),299-300.
  2. authors. title . Journal name. year of publication , volume issue , pg no.